MAT321761341 Matrix Scientific

Carbamimidoyl-acetic acid ethyl ester hydrochloride

  • MDL number MFCD06797615
  • Cas Number : 57508-48-2
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-108
Country of Origin USA
Product Link http://www.matrixscientific.com/020077.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/020077.html
Availability details for
Quantity
  • Availability
  • Add to Cart