MAT321761344 Matrix Scientific

4-[4-(Dimethylamino)benzylidene]-2-methyl-1,3-oxazol-5(4H)-one

  • MDL number MFCD00501269
  • Cas Number : 1787-23-1
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059706.html

Safety Information

MSDS http://www.matrixscientific.com/059706.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart