MAT321761350 Matrix Scientific

Ethyl 3-methyl-4,5,6,7-tetrahydro-1H-indole-2-carboxylate

  • MDL number MFCD00817001
  • Cas Number : 37945-37-2
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059712.html

Safety Information

MSDS http://www.matrixscientific.com/059712.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart