MAT321761352 Matrix Scientific

Ethyl 7-chloro-1-cyclopropyl-6-nitro-4-oxo-1,4-dihydro-3-quinolinecarboxylate

  • MDL number MFCD01088707
  • Cas Number : 127625-17-6
  • Molecular Weight : 336.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >280
Country of Origin USA
Product Link http://www.matrixscientific.com/059714.html

Safety Information

MSDS http://www.matrixscientific.com/059714.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart