MAT321761361 Matrix Scientific

17-Methyl-17-[(trimethylsilyl)oxy]estr-4-en-3-one

  • MDL number MFCD01746260
  • Cas Number : 77572-72-6
  • Molecular Weight : 360.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059723.html

Safety Information

MSDS http://www.matrixscientific.com/059723.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart