MAT321761364 Matrix Scientific

1-(5-Hydroxy-2-methyl-1-benzofuran-3-yl)ethanone

  • MDL number MFCD00464117
  • Cas Number : 28241-99-8
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238
Country of Origin USA
Product Link http://www.matrixscientific.com/059726.html

Safety Information

MSDS http://www.matrixscientific.com/059726.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart