MAT321761367 Matrix Scientific

2,3-Bis[(acetyloxy)methyl]-1-oxoquinoxalin--1-ium-4(1H)-olate

  • MDL number MFCD01106956
  • Cas Number : 10103-89-6
  • Molecular Weight : 306.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/059729.html

Safety Information

MSDS http://www.matrixscientific.com/059729.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart