MAT321761372 Matrix Scientific

3-[1-Ethyl-2,2-bis(4-methoxyphenyl)vinyl]-1H-indole

  • MDL number MFCD00847669
  • Cas Number : 354146-25-1
  • Molecular Weight : 383.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059734.html

Safety Information

MSDS http://www.matrixscientific.com/059734.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart