MAT321761373 Matrix Scientific

N-Phenyl-2-sulfanylacetamide

  • MDL number MFCD00014451
  • Cas Number : 4822-44-0
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-160
Country of Origin USA
Product Link http://www.matrixscientific.com/059735.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/059735.html
Availability details for
Quantity
  • Availability
  • Add to Cart