MAT321761374 Matrix Scientific

4-[2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethoxy]-4-oxobutanoic acid

  • MDL number MFCD00425441
  • Cas Number : 13182-87-1
  • Molecular Weight : 271.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109
Country of Origin USA
Product Link http://www.matrixscientific.com/059736.html

Safety Information

MSDS http://www.matrixscientific.com/059736.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart