MAT321761384 Matrix Scientific

Potassium S-(2,5-dihydroxyphenyl) thiosulfate

  • MDL number MFCD00810412
  • Cas Number : 88891-36-5
  • Molecular Weight : 260.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059746.html

Safety Information

MSDS http://www.matrixscientific.com/059746.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart