MAT321761385 Matrix Scientific

4-{[(2,2-Dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)-methyl]amino}butanoic acid

  • MDL number MFCD03067922
  • Cas Number : 1273578-10-1
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059747.html

Safety Information

MSDS http://www.matrixscientific.com/059747.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart