MAT321761387 Matrix Scientific

N,N-Diethyl-2-[(5-methyl-5H-pyridazino[3,4-b][1,4]benzoxazin-3-yl)oxy]ethanamine dihydrochloride

  • MDL number MFCD00378015
  • Cas Number : 55931-84-5
  • Molecular Weight : 387.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059749.html

Safety Information

MSDS http://www.matrixscientific.com/059749.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart