MAT321761392 Matrix Scientific

4,6-Dimethylthieno[2,3-b]pyridin-3(2H)-one

  • MDL number MFCD00854260
  • Cas Number : 55023-32-0
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-141
Country of Origin USA
Product Link http://www.matrixscientific.com/059671.html

Safety Information

MSDS http://www.matrixscientific.com/059671.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart