MAT321761393 Matrix Scientific

Magnesium bis(2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate)

  • MDL number MFCD00039120
  • Cas Number : 34717-03-8
  • Molecular Weight : 334.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/059672.html

Safety Information

MSDS http://www.matrixscientific.com/059672.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart