MAT321761396 Matrix Scientific

Methyl [4-(aminosulfonyl)phenyl]carbamate

  • MDL number MFCD00588271
  • Cas Number : 14070-56-5
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059758.html

Safety Information

MSDS http://www.matrixscientific.com/059758.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart