MAT321761397 Matrix Scientific

4-Nitrophenyl dihydrogen phosphate, sodium salt

  • MDL number MFCD18909627
  • Cas Number : 54306-27-3
  • Molecular Weight : 241.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059675.html

Safety Information

MSDS http://www.matrixscientific.com/059675.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart