MAT321761398 Matrix Scientific

Methyl 5-(4-chlorobenzylidene)-2,4-dioxo-1,3-thiazolidine-3-carboxylate

  • MDL number MFCD18800415
  • Cas Number : 181465-11-2
  • Molecular Weight : 297.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059760.html

Safety Information

MSDS http://www.matrixscientific.com/059760.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart