MAT321761410 Matrix Scientific

[3-(1-Phenylpropan-2-yl)-1,2,3-oxadiazol-3-ium-5-yl]azanide

  • MDL number MFCD00868768
  • Cas Number : 22293-47-6
  • Molecular Weight : 203.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059772.html

Safety Information

MSDS http://www.matrixscientific.com/059772.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart