MAT321761411 Matrix Scientific

5-Amino-3-(1-methyl-2-phenylethyl)-2,3-dihydro-1,2,3-oxadiazol-2-ium chloride

  • MDL number MFCD18800418
  • Cas Number : 3441-64-3
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059773.html

Safety Information

MSDS http://www.matrixscientific.com/059773.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart