MAT321761412 Matrix Scientific

3-Hexene-3,4-diylbis(4,1-phenylene)bis[dihydrogen (phosphate)]

  • MDL number MFCD00867397
  • Cas Number : 522-40-7
  • Molecular Weight : 428.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-206
Country of Origin USA
Product Link http://www.matrixscientific.com/059774.html

Safety Information

MSDS http://www.matrixscientific.com/059774.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart