MAT321761413 Matrix Scientific

3-{[2-(Hydroxymethyl)-2-phenylbutanoyl]oxy}-8,8-dimethyl-8-azoniabicyclo[3.2.1]octane iodide

  • MDL number MFCD18800419
  • Cas Number : 129109-88-2
  • Molecular Weight : 459.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/059775.html

Safety Information

MSDS http://www.matrixscientific.com/059775.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart