MAT321761415 Matrix Scientific

2-Hydroxy-5-[(4-{[(6-methoxy-3-pyridazinyl)amino]-sulfonyl}phenyl)diazenyl]benzoic acid

  • MDL number MFCD00864769
  • Cas Number : 22933-72-8
  • Molecular Weight : 429.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059777.html

Safety Information

MSDS http://www.matrixscientific.com/059777.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart