MAT321761417 Matrix Scientific

3-[2-(Tritylamino)ethyl]-1H-indol-5-ol

  • MDL number MFCD00337278
  • Cas Number : 19621-28-4
  • Molecular Weight : 418.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-110
Country of Origin USA
Product Link http://www.matrixscientific.com/059779.html

Safety Information

MSDS http://www.matrixscientific.com/059779.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart