MAT329179482 Matrix Scientific

2-Cyclopropyl-5-fluoropyrimidine-4,6-diol

  • MDL number MFCD19105237
  • Cas Number : 617715-97-6
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/059837.html

Safety Information

MSDS http://www.matrixscientific.com/059837.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart