MAT329179494 Matrix Scientific

Ethyl 5-ethoxy-1H-indole-2-carboxylate

  • MDL number MFCD03783503
  • Cas Number : 16382-17-5
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/059830.html

Safety Information

MSDS http://www.matrixscientific.com/059830.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart