MAT333115254 Matrix Scientific

1-Isoquinolin-5-ylmethanamine dihydrochloride

  • MDL number MFCD19442199
  • Cas Number : 58123-58-3
  • Molecular Weight : 231.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037474.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037474.html
Availability details for
Quantity
  • Availability
  • Add to Cart