MAT333115473 Matrix Scientific

[2-(Methylsulfinyl)ethyl]amine hydrobromide

  • MDL number MFCD16036096
  • Cas Number : 49773-19-5
  • Molecular Weight : 188.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060275.html

Safety Information

MSDS http://www.matrixscientific.com/060275.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart