MAT333115509 Matrix Scientific

N-(2-Thienylmethyl)butan-1-amine

  • MDL number MFCD05668044
  • Cas Number : 58924-53-1
  • Molecular Weight : 169.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/060311.html

Safety Information

MSDS http://www.matrixscientific.com/060311.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart