MAT333115548 Matrix Scientific

[(1-Cyclopropylpyrrolidin-3-yl)methyl]aminehydrochloride

  • MDL number MFCD13857459
  • Cas Number : 1017474-07-5
  • Molecular Weight : 176.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060350.html

Safety Information

MSDS http://www.matrixscientific.com/060350.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart