MAT333115699 Matrix Scientific

1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride

  • MDL number MFCD16039347
  • Cas Number : 1242339-08-7
  • Molecular Weight : 210.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060501.html

Safety Information

MSDS http://www.matrixscientific.com/060501.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart