MAT333115830 Matrix Scientific

(1-Imidazo[1,2-a]pyridin-2-ylethyl)aminedihydrochloride

  • MDL number MFCD18071251
  • Cas Number : 1266689-40-0
  • Molecular Weight : 234.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060632.html

Safety Information

MSDS http://www.matrixscientific.com/060632.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart