MAT333115875 Matrix Scientific

{4-[(Methylsulfonyl)methyl]phenyl}aminehydrochloride

  • MDL number MFCD18071261
  • Cas Number : 24176-70-3
  • Molecular Weight : 221.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060677.html

Safety Information

MSDS http://www.matrixscientific.com/060677.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart