MAT333115884 Matrix Scientific

3-Pyrrolidin-2-ylisoxazole hydrochloride

  • MDL number MFCD18071334
  • Cas Number : 1332528-62-7
  • Molecular Weight : 174.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060686.html

Safety Information

MSDS http://www.matrixscientific.com/060686.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart