MAT333115891 Matrix Scientific

3-(4-Methylpiperazin-1-yl)propan-1-oldihydrochloride

  • MDL number MFCD11841273
  • Cas Number : 82980-50-5
  • Molecular Weight : 231.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/060693.html

Safety Information

MSDS http://www.matrixscientific.com/060693.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart