MAT333115894 Matrix Scientific

[(3S)-3-Amino-3-carboxypropyl](dimethyl)-sulfonium iodide

  • MDL number MFCD00050272
  • Cas Number : 3493-11-6
  • Molecular Weight : 291.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Country of Origin USA
Product Link http://www.matrixscientific.com/060696.html

Safety Information

MSDS http://www.matrixscientific.com/060696.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart