MAT333115943 Matrix Scientific

N-(4-Fluorobenzyl)cyclopropanamine hydrochloride

  • MDL number MFCD07110865
  • Cas Number : 473732-87-5
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060745.html

Safety Information

MSDS http://www.matrixscientific.com/060745.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart