MAT333115988 Matrix Scientific

[(5-Methyl-1,3-thiazol-2-yl)methyl]aminedihydrochloride

  • MDL number MFCD18071410
  • Cas Number : 921091-08-9
  • Molecular Weight : 201.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060790.html

Safety Information

MSDS http://www.matrixscientific.com/060790.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart