MAT333116044 Matrix Scientific

[(1,2-Dimethyl-1H-imidazol-5-yl)methyl]aminedihydrochloride

  • MDL number MFCD18071367
  • Cas Number : 87786-09-2
  • Molecular Weight : 198.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/060846.html

Safety Information

MSDS http://www.matrixscientific.com/060846.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart