MAT333116075 Matrix Scientific

1-(1-Adamantyl)-4-methylpyridinium bromide

  • MDL number MFCD00168148
  • Cas Number : 20497-90-9
  • Molecular Weight : 308.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-189
Country of Origin USA
Product Link http://www.matrixscientific.com/060877.html

Safety Information

MSDS http://www.matrixscientific.com/060877.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart