MAT333116086 Matrix Scientific

(4-Chloro-3-nitrobenzyl)amine hydrochloride

  • MDL number MFCD06797911
  • Cas Number : 116599-39-4
  • Molecular Weight : 223.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/060888.html

Safety Information

MSDS http://www.matrixscientific.com/060888.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart