MAT333116150 Matrix Scientific

Potassium (methoxycarbonothioyl)sulfide

  • MDL number MFCD01880294
  • Cas Number : 34520-64-4
  • Molecular Weight : 146.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/060952.html

Safety Information

MSDS http://www.matrixscientific.com/060952.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart