MAT333116220 Matrix Scientific

2-(Piperidin-4-yloxy)benzoic acid hydrochloride

  • MDL number MFCD19103486
  • Cas Number : 1243250-02-3
  • Molecular Weight : 257.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061025.html

Safety Information

MSDS http://www.matrixscientific.com/061025.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart