MAT333116230 Matrix Scientific

[2-(1-Methylpiperidin-4-yl)ethyl]aminedihydrochloride

  • MDL number MFCD09455608
  • Cas Number : 20845-38-9
  • Molecular Weight : 215.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061035.html

Safety Information

MSDS http://www.matrixscientific.com/061035.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart