MAT333116283 Matrix Scientific

[2-(4-Ethylphenoxy)ethyl]amine

  • MDL number MFCD06654518
  • Cas Number : 67333-08-8
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061103.html

Safety Information

MSDS http://www.matrixscientific.com/061103.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart