MAT333116329 Matrix Scientific

(1-Adamantylthio)acetic acid

  • MDL number MFCD01821191
  • Cas Number : 95769-28-1
  • Molecular Weight : 226.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061149.html

Safety Information

MSDS http://www.matrixscientific.com/061149.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart