MAT333116349 Matrix Scientific

4-(2-Aminoethoxy)-3-methoxybenzoic acid

  • MDL number MFCD10039564
  • Cas Number : 1011408-00-6
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061169.html

Safety Information

MSDS http://www.matrixscientific.com/061169.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart