MAT333116351 Matrix Scientific

5-Methyl-10,11-dihydro-5H-dibenzo[b,f]azepine-2-carbaldehyde

  • MDL number MFCD06619352
  • Cas Number : 6487-67-8
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-93
Country of Origin USA
Product Link http://www.matrixscientific.com/061171.html

Safety Information

MSDS http://www.matrixscientific.com/061171.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart