MAT333116386 Matrix Scientific

1-(2-Aminophenyl)pyrrolidin-2-one

  • MDL number MFCD07339531
  • Cas Number : 14453-65-7
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061206.html

Safety Information

MSDS http://www.matrixscientific.com/061206.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart