MAT333116436 Matrix Scientific

(2E)-3-(2-Furyl)-1-phenylprop-2-en-1-one

  • MDL number MFCD00136235
  • Cas Number : 717-21-5
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/061256.html

Safety Information

MSDS http://www.matrixscientific.com/061256.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart